Learn More
ACROBiosystems Human Claudin-4 Protein, His,Twin-Strep Tag (Detergent)
Catalog No. 502332078
Encompass_Preferred
Human Claudin-4 Protein, His,Twin-Strep Tag (CL4-H5587) is expressed from Baculovirus-Insect cells. It contains AA Met 1 - Val 209 (Accession # O14493-1).
Catalog No. 50-233-2078 Supplier ACROBiosystems Supplier No. CL4H558720UG
Human Claudin-4 Protein, His,Twin-Strep Tag (CL4-H5587) is expressed from Baculovirus-Insect cells. It contains AA Met 1 - Val 209 (Accession # O14493-1).