Learn More
Biomatik Corporation Human Claudin 5 (CLDN5) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12516
Encompass_Preferred
Human Claudin 5 (CLDN5) Mini sample ELISA Kit for detecting CLDN5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00501. Gene ID: 7122. Target Synonyms: AWAL; BEC1; CPETRL1; TMVCF; Transmembrane Protein Deleted In Velocardiofacial Syndrome; Transmembrane protein deleted in VCFS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4372