Learn More
ACROBiosystems Human Claudin-6 100ug
Catalog No. 502535250
Encompass_Preferred
Human Claudin-6 Full Length Protein (VLP) (CL6-H52P7) is expressed from human 293 cells (HEK293). It contains AA Met 1 -Val 220 (Accession # P56747).
Catalog No. 50-253-5250 Supplier ACROBiosystems Supplier No. CL6H52P7100UG
Human Claudin-6 Full Length Protein (VLP) (CL6-H52P7) is expressed from human 293 cells (HEK293). It contains AA Met 1 -Val 220 (Accession # P56747).