Learn More
ACROBiosystems Human Claudin-6 20ug
Catalog No. 502535249
Encompass_Preferred
Human Claudin-6 Full Length Protein (VLP) (CL6-H52P7) is expressed from human 293 cells (HEK293). It contains AA Met 1 -Val 220 (Accession # P56747).
Catalog No. 50-253-5249 Supplier ACROBiosystems Supplier No. CL6H52P720UG
Human Claudin-6 Full Length Protein (VLP) (CL6-H52P7) is expressed from human 293 cells (HEK293). It contains AA Met 1 -Val 220 (Accession # P56747).