Learn More
Biomatik Corporation Human Claudin 6 (CLDN6) ELISA Kit
Supplier:  Biomatik Corporation EKU03215
Encompass_Preferred
Human Claudin 6 (CLDN6) ELISA Kit for detecting CLDN6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56747. Gene ID: 9074. Target Synonyms: Skullin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7587