Learn More
ACROBiosystems Human Claudin-9, His,Twin-Strep Tag (CL9-H5586) is expressed from Baculovirus-Insect cells. It contains AA Ala 2 - Val 217 (Accession # O95484-1).
Catalog No. 502254114
Encompass_Preferred
Claudin-9 belongs to the claudin family. Claudins constitute integral membrane proteins responsible for solute and electrolyte permeability of the tight junction that serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets. Tight junctions also play a critical role in maintaining cell polarity and signal transductions. Claudin-9 creates charge specific channels in the paracellular space, plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity, is required to preserve sensory cells in the hearing organ because claudin-9-defective tight junctions fail to shield the basolateral side of hair cells from the K+-rich endolymph. Its ion barrier function is essential in the cochlea, but appears to be dispensable in other organs. Is one of the entry cofactors for hepatitis C virus; it enables HCV entry into target cell
This item is not returnable. View return policy
Catalog No. 50-225-4114 Supplier ACROBiosystems Supplier No. CL9H558620UG
Claudin-9 belongs to the claudin family. Claudins constitute integral membrane proteins responsible for solute and electrolyte permeability of the tight junction that serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets. Tight junctions also play a critical role in maintaining cell polarity and signal transductions. Claudin-9 creates charge specific channels in the paracellular space, plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity, is required to preserve sensory cells in the hearing organ because claudin-9-defective tight junctions fail to shield the basolateral side of hair cells from the K+-rich endolymph. Its ion barrier function is essential in the cochlea, but appears to be dispensable in other organs. Is one of the entry cofactors for hepatitis C virus; it enables HCV entry into target cell