Learn More
Sino Biological Anti-NNT1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10937MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NNT1 (rh NNT1; Catalog#10937-H01H; NP_037378.1; Leu28-Phe225). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5047