Learn More
Sino Biological Anti-Claudin-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200156T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Claudin-1/CLDN1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3971