Learn More
Sino Biological Human CLDN1 PAb 100UL 101363-T32-100
Supplier:  Sino Biological 101363T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Claudin-1 / CLDN1, and purified by antigen affinity chromatography.
Catalog No. 50-241-0074