Learn More
Sino Biological Anti-CLDN12 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200221T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CLDN12 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3969