Learn More
Sino Biological Anti-Claudin18 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200116T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Claudin18 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3975