Learn More
Sino Biological Human CLDN18 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 109916T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Claudin18, and purified by antigen affinity chromatography.
Catalog No. 50-237-2954