Learn More
Sino Biological Recombinant Human Claudin-4/CLDN4 Protein (VLP, Full-length, Flag Tag)
Catalog No. 502993172
Encompass_Preferred
A DNA sequence encoding the Human CLDN4-VLP (Full Length) (O14493) (Met1-Val209) was fused with a flag tag at the C-terminus.
Catalog No. 50-299-3172 Supplier Sino Biological Supplier No. 16358H25H100
A DNA sequence encoding the Human CLDN4-VLP (Full Length) (O14493) (Met1-Val209) was fused with a flag tag at the C-terminus.