Learn More
Biomatik Corporation Human Cleavage And Polyadenylation Specific Factor 1 (CPSF1) ELISA Kit
Supplier:  Biomatik Corporation EKU03218
Encompass_Preferred
Human Cleavage And Polyadenylation Specific Factor 1 (CPSF1) ELISA Kit for detecting CPSF1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q10570. Gene ID: 29894. Target Synonyms: CPSF160; Cleavage and polyadenylation specificity factor 160 kDa subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6304