Learn More
ACROBiosystems Human CLEC-2, His Tag (CL2-H5247) is expressed from human 293 cells (HEK293). It contains AA Ser 55 - Pro 229 (Accession # Q9P126-1).
Supplier:  ACROBiosystems CL2H52471MG
Encompass_Preferred
C-type lectin-like receptor 2 (CLEC-2) is a hemITAM containing receptor that is highly expressed on platelets and regulates many of the non-classical functions of these cells, including in inflammation and infection.
Catalog No. 50-225-3826