Learn More
Sino Biological Anti-CLEC10A/CD301 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201405T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CLEC10A fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3945