Learn More
ACROBiosystems Human CLEC12A 25tests
Catalog No. 502535237
Encompass_Preferred
PE-Labeled Human CLEC12A Protein His Tag (CLA-HP245) is produced via conjugation of PE to Human CLEC12A Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CLEC12A Protein His Tag is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2).
Catalog No. 50-253-5237 Supplier ACROBiosystems Supplier No. CLAHP24525TESTS
PE-Labeled Human CLEC12A Protein His Tag (CLA-HP245) is produced via conjugation of PE to Human CLEC12A Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CLEC12A Protein His Tag is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2).