Learn More
ACROBiosystems APC-Labeled Human CLEC12A / MICL / CLL-1 Protein, His Tag (Site-specific conjugation)
Catalog No. 502331736
Encompass_Preferred
APC-Labeled Human CLEC12A, His Tag (CLA-HA248) is produced via conjugation of APC to Human CLEC12A, His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human CLEC12A, His Tag is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # NP_612210.4).
Catalog No. 50-233-1736 Supplier ACROBiosystems Supplier No. CLAHA24825TESTS
APC-Labeled Human CLEC12A, His Tag (CLA-HA248) is produced via conjugation of APC to Human CLEC12A, His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human CLEC12A, His Tag is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # NP_612210.4).