Learn More
Sino Biological Recombinant Human CLL-1/CLEC12A Protein (AVI & Fc Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11896H42HB20
Encompass_Preferred
A DNA sequence encoding the human CLEC12A (EAW96132.1) (His75-Ala275) was expressed with a N-terminal AVI tag followed by a Fc region of human IgG1 tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed..
Catalog No. 50-241-0675