Learn More
Sino Biological Human CLEC1B PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10976T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CLEC1B / CLEC2 (rh CLEC1B / CLEC2; Catalog#10976-H07H; NP_057593.3; Gln58-Pro229). CLEC1B / CLEC2 specific IgG was purified by Human CLEC1B / CLEC2 affinity chromatography.
Catalog No. 50-237-3708