Learn More
BIOHIPPO Human CLEC3B Microsample
SDP
Catalog No. 5000313807
Encompass
Human CLEC3B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CLEC3B in Plasma It provides a detection range of 0 19-12 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CLEC3B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CLEC3B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CLEC3B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13807 Supplier BIOHIPPO Supplier No. BHE15211205
Human CLEC3B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CLEC3B in Plasma It provides a detection range of 0 19-12 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CLEC3B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CLEC3B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CLEC3B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color