Learn More
Sino Biological Human CLEC4A ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502370946
Encompass_Preferred
Each vial contains 45 ng of recombinant CLEC4A. Reconstitute standard powder with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.
Catalog No. 50-237-0946 Supplier Sino Biological Supplier No. SEK114765
Each vial contains 45 ng of recombinant CLEC4A. Reconstitute standard powder with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.