Learn More
Sino Biological Recombinant Anti-DICR/CLEC4A Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11476R007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human DCIR / CLEC4A / CLECSF6 (rh DCIR / CLEC4A / CLECSF6; Catalog#11476-H07H; NP_057268.1; Gln 70-Leu 237).
Catalog No. 50-241-1890