Learn More
Sino Biological Anti-DICR/CLEC4A Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11476MM08P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DCIR / CLEC4A / CLECSF6 (rh DCIR / CLEC4A / CLECSF6; Catalog#11476-H07H; NP_057268.1; Gln 70-Leu 237) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8441