Learn More
Sino Biological Anti-DICR/CLEC4A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201490T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CLEC4A fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2528