Learn More
Sino Biological Anti-DICR/CLEC4A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11476RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CLEC4A (rh CLEC4A; Catalog#11476-H07H; NP_057268.1; Gln 70-Leu 237). CLEC4A specific IgG was purified by human CLEC4A affinity chromatography.
Catalog No. 50-240-8074