Learn More
Sino Biological Recombinant Human CLEC4C Protein (ECD, hFc Tag)
Catalog No. 502993582
Encompass_Preferred
A DNA sequence encoding the Human CLEC4C (NP_569708.1) (Asn45-Ile213) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-299-3582 Supplier Sino Biological Supplier No. 19905H01H100
A DNA sequence encoding the Human CLEC4C (NP_569708.1) (Asn45-Ile213) was expressed with the Fc region of human IgG1 at the N-terminus.