Learn More
ACROBiosystems Human CLEC4C, Fc Tag (CLC-H5254) is expressed from human 293 cells (HEK293). It contains AA Asn 45 - Ile 213 (Accession # Q8WTT0-1).
Supplier:  ACROBiosystems CLCH5254100UG
Encompass_Preferred
CLEC4C (C-type lectin domain family 4 member C), also known as BDCA2, CLECSF11, CLECSF7, DLEC and CD303. Lectin-type cell surface receptor which may play a role in antigen capturing by dendritic cell. Specifically recognizes non-sialylated galactose-terminated biantennary glycans containing the trisaccharide epitope Gal(beta1-3/4)GlcNAc(beta1-2)Man. Binds to serum IgG. Efficiently targets ligand into antigen-processing and peptide-loading compartments for presentation to T-cells. May mediate potent inhibition of induction of IFN-alpha/beta expression in plasmacytoid dendritic cells. May act as a signaling receptor that activates protein-tyrosine kinases and mobilizes intracellular calcium.
Catalog No. 50-223-1255