Learn More
Sino Biological Anti-CLEC4D Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11485RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CLEC4D (rh CLEC4D; Catalog#11485-H07H; NP_525126.2; Gly52-Asn215). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4707