Learn More
Sino Biological Recombinant Human Dectin-2/CLEC6A Protein (ECD, His Tag)
Supplier:  Sino Biological 10250H08H1
Encompass_Preferred
A DNA sequence encoding the Human CLEC6A (NP_001007034) (Thr42-Leu209) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5412