Learn More
BIOHIPPO Human CLL1 Microsample
SDP
Catalog No. 5000314368
Encompass
Human CLL1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CLL1 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 56 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CLL1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CLL1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CLL1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14368 Supplier BIOHIPPO Supplier No. BHE15211254
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Human CLL1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CLL1 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 56 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CLL1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CLL1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CLL1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color