Learn More
Sino Biological Anti-ASAM Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10794MM05P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ASAM / CLMP (rh ASAM / CLMP; Catalog#10794-H08H; NP_079045.1; Met1-Met233) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1927