Learn More
Sino Biological Anti-CLPS Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13631RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CLPS / Colipase (rh CLPS; Catalog#13631-H08B; P04118; Met1-Gln112). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4204