Learn More
Sino Biological Human CLU MAb 25TS (Sino Biological)
Supplier:  Sino Biological 11297R013F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Clusterin / Apolipoprotein J / Apo-J (rh Clusterin / Apolipoprotein J / Apo-J; Catalog#11297-H08H; NP_001822.2; Met1-Glu501) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-237-1526