Learn More
Sino Biological Anti-Clusterin Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11297MM01F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Clusterin / Apolipoprotein J / Apo-J (rh Clusterin / Apolipoprotein J / Apo-J; Catalog#11297-H08H; NP_001822.2; Met1-Glu501) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8880