Learn More
Sino Biological Recombinant Anti-Clusterin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11297R012100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Clusterin / Apolipoprotein J / Apo-J (rh Clusterin / Apolipoprotein J / Apo-J; Catalog#11297-H08H; NP_001822.2; Met 1-Glu 501).
Catalog No. 50-240-9185