Learn More
Sino Biological Human CLU MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11297R210100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Clusterin / Apolipoprotein J / Apo-J / CLU (rh Clusterin / Apolipoprotein J / Apo-J / CLU; Catalog#11297-H08H; NP_001822.2; Met1-Glu501).
Catalog No. 50-237-2366