Learn More
Sino Biological Recombinant Human Clusterin Protein (sCLU, His Tag)
Supplier:  Sino Biological 11297H08H1100
Encompass_Preferred
A DNA sequence encoding the Human CLU (NP_001822.3) (Met1-Glu449) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-241-1463