Learn More
American Research Products Inc Human Cluster of differentiati
Catalog No. 5000329750
Encompass
Human Cluster of differentiation 8 CD8 ELISA Kit from CUSABIO Detection Range 62 5 U/mL-4000 U/mL Sensitivity 15 6 U/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29750 Supplier American Research Products Inc Supplier No. CSBE12707H
Human Cluster of differentiation 8 CD8 ELISA Kit from CUSABIO Detection Range 62 5 U/mL-4000 U/mL Sensitivity 15 6 U/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich