Learn More
American Research Products Inc Human cluster of differentiati
Catalog No. 5000330250
Encompass
Human cluster of differentiation 63 CD63 ELISA Kit from CUSABIO Detection Range 47pg/mL-3000pg/mL Sensitivity 23 5pg/ml Sample serum plasma cell culture supernates urine saliva tissue homogenates Method Sandwich
Catalog No. 50-003-30250 Supplier American Research Products Inc Supplier No. CSBE14107HIS
Human cluster of differentiation 63 CD63 ELISA Kit from CUSABIO Detection Range 47pg/mL-3000pg/mL Sensitivity 23 5pg/ml Sample serum plasma cell culture supernates urine saliva tissue homogenates Method Sandwich