Learn More
Biomatik Corporation Human Cluster Of Differentiation 1a (CD1a) ELISA Kit
Supplier:  Biomatik Corporation EKU12291
Encompass_Preferred
Human Cluster Of Differentiation 1a (CD1a) ELISA Kit for detecting CD1a in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06126. Gene ID: 909. Target Synonyms: CD1-A; CD1; FCB6; HTA1; R4; T6; T-cell surface antigen T6/Leu-6; hTa1 thymocyte antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5806