Learn More
Biomatik Corporation Human Cluster Of Differentiation 226 (CD226) ELISA Kit
Supplier:  Biomatik Corporation EKU03232
Encompass_Preferred
Human Cluster Of Differentiation 226 (CD226) ELISA Kit for detecting CD226 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15762. Gene ID: 10666. Target Synonyms: DNAM1; PTA1; TLiSA1; DNAX accessory molecule 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7084