Learn More
Biomatik Corporation Human Cluster Of Differentiation 99 (CD99) ELISA Kit
Supplier:  Biomatik Corporation EKU03248
Encompass_Preferred
Human Cluster Of Differentiation 99 (CD99) ELISA Kit for detecting CD99 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14209. Gene ID: 4267. Target Synonyms: MIC2X; MIC2Y; MIC2; Antigen Identified By Monoclonal Antibodies 12E7,F21 And O13; E2 antigen; T-cell surface glycoprotein E2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7088