Learn More
Biomatik Corporation Human Cluster Of Differentiation 30 Ligand (CD30L) ELISA Kit
Supplier:  Biomatik Corporation EKU03235
Encompass_Preferred
Human Cluster Of Differentiation 30 Ligand (CD30L) ELISA Kit for detecting CD30L in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 34pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32971. Gene ID: 944. Target Synonyms: CD153; TNFSF8; CD30-L; Tumor Necrosis Factor Ligand Superfamily Member 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9054