Learn More
Raybiotech Inc Human Clusterin ELISA Kit
Supplier:  Raybiotech Inc ELHCLUSTERIN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 15 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detect human Clusterin. Other species not determined.
Catalog No. 50-194-9130