Learn More
CPC Scientific H-Asn-Leu-Val-Pro-Met-Val-Ala-Thr-Val-OH 1MG
SDP
Supplier:  CPC Scientific HCMV001A
Encompass
This peptide is the immunodominant epitope derived from the human cytomegalovirus (HCMV) matrix protein pp65.SEQUENCE: H-Asn-Leu-Val-Pro-Met-Val-Ala-Thr-Val-OHONE-LETTER SEQUENCE: NLVPMVATVMOLECULAR FORMULA: C42H74N10O12S1MOLECULAR WEIGHT: 943.18STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [153045-21-7]SYNONYMS: pp65 (495-503) (human cytomegalovirus), 65 kDa Lower Matrix Phosphoprotein (495-503) (human cytomegalovirus)RESEARCH AREA: AntimicrobialREFERENCES:J.W.Gratama et al., Blood, 98, 1358 (2001)S.Szmania et al., Blood, 98, 505 (2001)K.S.Lang et al., Am. J. Physiol. Cell Physiol., 284, C200 (2003)H.Hebart et al., Exp. Hematol., 31, 966 (2003)P.S.Rohrlich et al., Hum. Immunol., 65, 514 (2004)SKU(s): HCMV-001A, HCMV-001B
Catalog No. 50-210-9418