Learn More
Sino Biological Recombinant Human CNDP2 Protein (His Tag)
Supplier:  Sino Biological 10168H08B50
Encompass_Preferred
A DNA sequence encoding the human CNDP2 (CAC69883.1) (Met 1-Asp 475) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6734