Learn More
BIOHIPPO Human CNP Microsample
SDP
Catalog No. 5000315212
Encompass
Human CNP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CNP in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 25 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CNP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CNP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CNP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15212 Supplier BIOHIPPO Supplier No. BHE15210745
Human CNP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CNP in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 25 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CNP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CNP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CNP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color