Learn More
Proteintech Group Inc Human CNPY2 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0007396T
Encompass_Preferred
CNPY2, also named as MSAP, TMEM4 and ZSIG9, is a positive regulator of neurite outgrowth (PMID: 12826659). It interacts with the ezrin-radixin-moesin (ERM)-like myosin regulatory light chain-interacting protein (MIR), and the two proteins are co-localized in cell lines and in primary neurons. CNPY2 is a HIF-1α regulated, secreted angiogenic growth factor that promotes smooth muscle cells migration, proliferation and tissue revascularization (PMID: 25589425). CNPY2 enhances cell spreading in fibroblasts and migration of rat C6 glioma cells (PMID: 15905959). CNPY2 was also shown to influence the expression of low-density lipoprotein receptor and FGF21 (PMID: 22378787). This kit is used to quantify CNPY2 level in vivo.
Catalog No. 50-197-4039