Learn More
Sino Biological Anti-CNPY2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13771T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CNPY2 (rh CNPY2; Catalog#13771-H08H; Q9Y2B0-1; Met1-Ser178). CNPY2 specific IgG was purified by Human CNPY2 affinity chromatography.
Catalog No. 50-255-4191